Learn More
Enzo Life Sciences (+)-Bicuculline (250mg). CAS: 485-49-4
SDP
Supplier:  Enzo Life Sciences ALX550515M250
Encompass_Preferred
GABAA receptor antagonist. Alternative name: D-Bicuculline. Purity: ≥98% (HPLC). Appearance: Off-white to yellow powder. Solubility: Soluble in benzene, chloroform or ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-200-9558