Learn More
Selleck Chemical LLC (+)-Bicuculline 25mg 485-49-4
SDP
Supplier:  Selleck Chemical LLC S707125MG
Encompass_Preferred
(+)-Bicuculline is a competitive antagonist of GABAA receptors with IC50 of 2 ?M, also blocks Ca(2+)-activated potassium channels. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1579000