Learn More
Medchemexpress LLC HY-N0219 50mg Medchemexpress, (+)-Bicuculline CAS:485-49-4 Purity:>98%
SDP
Catalog No. 501933475
Encompass_Preferred
Medchemexpress, HY-N0219 50mg (+)-Bicuculline CAS:485-49-4 (+)-Bicuculline is a light-sensitive competitive antagonist of GABA-A receptor . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3475 Supplier Medchemexpress LLC Supplier No. HYN021950MG
Medchemexpress, HY-N0219 50mg (+)-Bicuculline CAS:485-49-4 (+)-Bicuculline is a light-sensitive competitive antagonist of GABA-A receptor . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.