Learn More
Enzo Life Sciences (+)-Bicuculline (50mg). CAS: 485-49-4
SDP
Catalog No. 502009557
Encompass_Preferred
GABAA receptor antagonist. Alternative name: D-Bicuculline. Purity: ≥98% (HPLC). Appearance: Off-white to yellow powder. Solubility: Soluble in benzene, chloroform or ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-200-9557 Supplier Enzo Life Sciences Supplier No. ALX550515M050
GABAA receptor antagonist. Alternative name: D-Bicuculline. Purity: ≥98% (HPLC). Appearance: Off-white to yellow powder. Solubility: Soluble in benzene, chloroform or ethyl acetate. Long Term Storage: -20°C.