Learn More
Enzo Life Sciences (-)-Bicuculline methiodide (50mg). CAS: 40709-69-1
SDP
Catalog No. 502010538
Encompass_Preferred
A water soluble analog of bicuculline, the classic GABAA antagonist. Purity: ≥98% (HPLC). Solubility: Soluble in water (10mg/ml) or DMSO (30mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0538 Supplier Enzo Life Sciences Supplier No. BMLEA1490050
A water soluble analog of bicuculline, the classic GABAA antagonist. Purity: ≥98% (HPLC). Solubility: Soluble in water (10mg/ml) or DMSO (30mg/ml). Long Term Storage: Ambient.