Learn More
Medchemexpress LLC HY-100783 10mg Medchemexpress, (-)-Bicuculline (methobromide) CAS:73604-30-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10078310MG
Encompass_Preferred
Medchemexpress, HY-100783 10mg (-)-Bicuculline (methobromide) CAS:73604-30-5 (-)-Bicuculline methobromide (l-Bicuculline methobromide) is a potent GABAA receptor antagonist. (-)-Bicuculline methobromide blocks afterhyperpolarizations (AHPs) mediated by Ca2+-activated K+ channels in various types of neurons. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0601