Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid, 1-methyl ester

Catalog No. PHB00015 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHB00015 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHB00015 Supplier Sigma Aldrich Supplier No. PHB00015100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-[2-(2-chloroethoxy)ethoxy]ethanol was used in the synthesis of 1-[3-(2-aminoethyl)-1h-indol-5-yloxy]-3,6-dioxa-8-mercaptooctane
  • It was also used in the synthesis of modified oligonucleotides bearing a disulfide or a thiol function at the 3α-end and other modifications at the 5α-end

Specifications

CAS 83249-10-9
MDL Number MFCD20638314
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.