Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bicyclo[2.2.1]hepta-2,5-diene

Catalog No. B338035ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B338035ML 5 mL
B33803500ML 500 mL
B33803100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B338035ML Supplier Sigma Aldrich Supplier No. B338035ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Intermediate in prostaglandin synthesis.

Specifications

CAS 121-46-0
Density 0.906 g/mL (at 25°C)
Boiling Point 89°C
Molecular Formula C7H8
Refractive Index n20/D 1.470 (literature)
Linear Formula C7H8
Quantity 5 mL
MDL Number MFCD00082301
Synonym 2,5-Norbornadiene; NBD
Molecular Weight (g/mol) 92.14
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.