Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[2.2.1]hepta-2,5-diene, Stabilized 96%, Thermo Scientific™

Catalog No. AC106240050
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106240050 5 mL
AC106240500 50 mL
AC106242500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC106240050 Supplier Thermo Scientific Chemicals Supplier No. 106240050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-46-0
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00082301
InChI Key SJYNFBVQFBRSIB-UHFFFAOYSA-N
Synonym 2,5-norbornadiene, bicyclo 2.2.1 hepta-2,5-diene, norbornadiene, unii-w9ztq75zus, bicyclo 2.2.1 heptadiene, 8,9,10-trinorborna-2,5-diene, w9ztq75zus, dicycloheptadiene
PubChem CID 8473
IUPAC Name bicyclo[2.2.1]hepta-2,5-diene
SMILES C1C2C=CC1C=C2

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4690 to 1.4710 (20°C, 589nm)
Quantity 5 mL
Merck Index 15, 6776
Stabilizer 0.05 to 0.25% BHT
Formula Weight 92.14
Percent Purity ≥97.0%
Physical Form Liquid
Chemical Name or Material Bicyclo[2.2.1]hepta-2, 5-diene, 96%
Show More Show Less
EINECSNumber 204-472-0
RTECSNumber RB6535000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.