Learn More
Medchemexpress LLC Bicyclo[2.2.1]heptane-1-carboxylic acid, 4-[2-[9-[(4-fluorophenyl)methyl]-5,6,8,9-tetrahydro... | 1354805-08-5 | 99.9% | 461.53 g/mol | C27H28FN3O3 | 5 MG
SDP
Catalog No. 5000221321
Encompass_Preferred
ONO-8430506 is a potent, orally bioavailable autotaxin (ATX/ENPP2) inhibitor used for biochemical assays and preclinical studies. It exhibits low-nanomolar enzymatic potency and has demonstrated in vivo activity in rodent tumor models, with established solubility and storage data for laboratory use.
  • Potent ATX inhibition with low-nanomolar IC50 values in enzymatic assays.
  • IC90 of 100 nM for ATX activity in mouse plasma.
  • Orally bioavailable and effective in preclinical tumor models.
  • Documented solubility in DMSO and multiple in vivo formulation options.
  • High supplied purity and specified storage conditions for stability.
Catalog No. 50-002-21321 Supplier Medchemexpress LLC Supplier No. HY1453445MG
ONO-8430506 is a potent, orally bioavailable autotaxin (ATX/ENPP2) inhibitor used for biochemical assays and preclinical studies. It exhibits low-nanomolar enzymatic potency and has demonstrated in vivo activity in rodent tumor models, with established solubility and storage data for laboratory use.
  • Potent ATX inhibition with low-nanomolar IC50 values in enzymatic assays.
  • IC90 of 100 nM for ATX activity in mouse plasma.
  • Orally bioavailable and effective in preclinical tumor models.
  • Documented solubility in DMSO and multiple in vivo formulation options.
  • High supplied purity and specified storage conditions for stability.