Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[3.2.1]octan-2-one 90-95%, Thermo Scientific™

Catalog No. AC152710010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC152710010 1 g
AC152710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152710010 Supplier Thermo Scientific Chemicals Supplier No. 152710010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5019-82-9
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
InChI Key SXFPXZSENHPCSH-UHFFFAOYSA-N
Synonym bicyclo 3.2.1 octan-2-one, bicyclo 3.2.1 octan-4-one, bicyclo 3,2,1 octan-2-one, bicyclo 3.2.1 octanone, 4-bicyclo 3.2.1 octanone, ghl.pd_mitscher_leg0.241, bicyclo 3.2.1 oct-2-one, bicyclo 3.2.1 octan-2-on
PubChem CID 107363
IUPAC Name bicyclo[3.2.1]octan-4-one
SMILES C1CC2CC1CCC2=O

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 118°C to 124°C
Color White
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 124.18
Percent Purity ≥90%
Physical Form Crystalline Powder
Chemical Name or Material Bicyclo[3.2.1]octan-2-one, 90-95%
EINECSNumber 225-704-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.