Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclohexane-4,4'-dione Monoethylene Ketal 98.0+%, TCI America™
SDP

Catalog No. B34055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34055G 5 g
B340525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34055G Supplier TCI America Supplier No. B34055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56309-94-5
Molecular Formula C14H22O3
Molecular Weight (g/mol) 238.327
MDL Number MFCD00809704
InChI Key ZNWLFTSPNBLXGL-UHFFFAOYSA-N
Synonym ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl
PubChem CID 810913
IUPAC Name 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one
SMILES C1CC(=O)CCC1C2CCC3(CC2)OCCO3

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 5 g
Formula Weight 238.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bicyclohexane-4,4′-dione Monoethylene Ketal
EINECSNumber (5)-6425
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.