Learn More
Medchemexpress LLC BICYCLOMYCIN BENZOATE 10MG
SDP
Catalog No. NC2256119
Encompass_Preferred
Medchemexpress,HY-101128 10mg Bicyclomycin benzoate FR2054 CAS:37134-40-0 Formula:C19H22N2O8 Bicyclomycin benzoate is an antibiotic exhibiting activity against a broad spectrum of Gram-negative bacteria and against the Gram-positive bacterium. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered
This item is not returnable. View return policy
Catalog No. NC2256119 Supplier Medchemexpress LLC Supplier No. HY10112810MG
Medchemexpress,HY-101128 10mg Bicyclomycin benzoate FR2054 CAS:37134-40-0 Formula:C19H22N2O8 Bicyclomycin benzoate is an antibiotic exhibiting activity against a broad spectrum of Gram-negative bacteria and against the Gram-positive bacterium. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered