Learn More
Medchemexpress LLC BICYCLOMYCIN BENZOATE 25MG
SDP
Supplier:  Medchemexpress LLC HY10112825MG
Encompass_Preferred
Medchemexpress,HY-101128 25mg Bicyclomycin benzoate FR2054 CAS:37134-40-0 Formula:C19H22N2O8 Bicyclomycin benzoate is an antibiotic exhibiting activity against a broad spectrum of Gram-negative bacteria and against the Gram-positive bacterium. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered
Catalog No. NC2256123