Learn More
R&D Systems™ BID Antibody Pair [HRP]

Description
Specifications
Specifications
| Assay Range | 7.8 pg/mL |
| Assay Sensitivity | 7.81-500 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Line | Novus |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human BID. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.