Learn More
Medchemexpress LLC BID Mouse His-GST 10ug
SDP
Catalog No. 5000181667
Encompass_Preferred
BID Protein a pro-apoptotic member of the Bcl-2 family is initially discovered through binding to both pro-apoptotic Bax and anti-apoptotic Bcl-2 BID is activated in the BCL-2-regulated or mitochondrial apoptosis pathway and acts as a switch between the extrinsic and intrinsic cell death pathways BID is susceptible to proteolytic cleavage by caspases calpains Granzyme B and cathepsins BID Protein Mouse (His-GST) is the recombinant mouse-derived BID protein expressed by E coli with N-His N-GST labeled tag
Catalog No. 50-001-81667 Supplier Medchemexpress LLC Supplier No. HYP7285210UG
BID Protein a pro-apoptotic member of the Bcl-2 family is initially discovered through binding to both pro-apoptotic Bax and anti-apoptotic Bcl-2 BID is activated in the BCL-2-regulated or mitochondrial apoptosis pathway and acts as a switch between the extrinsic and intrinsic cell death pathways BID is susceptible to proteolytic cleavage by caspases calpains Granzyme B and cathepsins BID Protein Mouse (His-GST) is the recombinant mouse-derived BID protein expressed by E coli with N-His N-GST labeled tag