Learn More
Sigma Aldrich ™-Bidime
Catalog No. 913200100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
500 mg
Catalog No. Quantity
91-320-0100MG 500 mg
1 options
Catalog No. 91-320-0100MG Supplier Sigma Aldrich Supplier No. 913200100MG
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
(R)-BIDIME is a P-chiral monophosphorus ligand used for the transition metal-catalyzed asymmetric Suzuki-Miyaura and hydroboration reactions.Synthesis of Chiral alpha-Amino Tertiary Boronic Esters by Enantioselective Hydroboration of alpha-ArylenamidesSold in collaboration with Zejun Pharmaceuticals
Specifications
CAS 3482-9-6-9
Quantity 500 mg