Learn More
National Diagnostics BIEBRICH SCARLET 25 GR
SDP
Catalog No. 5089990156
Encompass
Biebrich Scarlet, Plasma stain substituted for Acid Fuchsin in Masson's trichrome, 25 GR
Catalog No. 50-899-90156 Supplier National Diagnostics Supplier No. HS50625GRM
Biebrich Scarlet, Plasma stain substituted for Acid Fuchsin in Masson's trichrome, 25 GR