Learn More
Scytek Laboratories BIEBRICH SCARLET/ACID FUCHS1L
SDP
Catalog No. NC2035349
Encompass_Preferred
Biebrich Scarlet/Acid Fuchsin Solution, 1 L. Used in Masson's Trichrome.
Catalog No. NC2035349 Supplier Scytek Laboratories Supplier No. BSU999
Biebrich Scarlet/Acid Fuchsin Solution, 1 L. Used in Masson's Trichrome.