Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Biebrich Scarlet, Water Soluble 25 GR
SDP

Catalog No. 50980196
Encompass_Preferred

(Bonceau BS; Acid Red 66; Croceine Scarlet; Double Scarlet BSF)
CAS #4196-99-0 C₂₂H₁₄N₄Na₂O₇S₂ F.W. 556.4 9
Solubility: 5.0% Water; 0.25% Alcohol; 1.5% Cellosolve; 1.5% Glycol; 0.0% Xylene

Used with picric acid/aniline blue for staining collagen, recticulum, muscle, and plasma. Also Luna's method for erythrocytes & eosinophil granules and Guard's method for sex chromatin and nuclear chromatin. Biebrich Scarlet, water soluble is also known as Bonceau BS; Acid Red 66; Croceine Scarlet and Double Scarlet BSF.

Catalog No. 50-980-196 Supplier Electron Microscopy Sciences Supplier No. 11265
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Bonceau BS; Acid Red 66; Croceine Scarlet; Double Scarlet BSF)
CAS #4196-99-0 C₂₂H₁₄N₄Na₂O₇S₂ F.W. 556.4 9
Solubility: 5.0% Water; 0.25% Alcohol; 1.5% Cellosolve; 1.5% Glycol; 0.0% Xylene

Used with picric acid/aniline blue for staining collagen, recticulum, muscle, and plasma. Also Luna's method for erythrocytes & eosinophil granules and Guard's method for sex chromatin and nuclear chromatin. Biebrich Scarlet, water soluble is also known as Bonceau BS; Acid Red 66; Croceine Scarlet and Double Scarlet BSF.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.