Learn More
Newcomer Supply Bielschowsky, Lester-King Modified Stain Kit, 250 ml kit
SDP
Catalog No. NC1381156
Encompass
Newcomer Supply Bielschowsky, Lester King Modified Stain Kit procedure is used to demonstrate nerve fibers, neurofibrils/tangles, senile plaques and axons.  This stain can be instrumental in the diagnosis of Alzheimer’s disease and other neurological disorders.
Catalog No. NC1381156 Supplier Newcomer Supply Supplier No. 9154A
Newcomer Supply Bielschowsky, Lester King Modified Stain Kit procedure is used to demonstrate nerve fibers, neurofibrils/tangles, senile plaques and axons.  This stain can be instrumental in the diagnosis of Alzheimer’s disease and other neurological disorders.