Learn More
Medchemexpress LLC Bifemelane hydrochloride (MCI-2016) | 62232-46-6 | 99.9% | 10 MM * 1 ML
SDP
Catalog No. 5000412035
Encompass_Preferred
Bifemelane hydrochloride (MCI-2016) is a potent, selective, and competitive inhibitor of monoamine oxidase A (MAO-A), with a Ki of 4.20 μM. It also inhibits MAO-B noncompetitively with a Ki of 46.0 μM. Bifemelane hydrochloride has antidepressant activity and is used for research into cognitive and emotional disturbances related to cerebrovascular disease.
  • Potent, selective, and competitive inhibitor of monoamine oxidase A (MAO-A)
  • Inhibits MAO-B noncompetitively
  • Antidepressant activity
  • Used for research into cognitive and emotional disturbances related to cerebrovascular disease
Catalog No. 50-004-12035 Supplier Medchemexpress LLC Supplier No. HYB1558A10MM/1ML
Bifemelane hydrochloride (MCI-2016) is a potent, selective, and competitive inhibitor of monoamine oxidase A (MAO-A), with a Ki of 4.20 μM. It also inhibits MAO-B noncompetitively with a Ki of 46.0 μM. Bifemelane hydrochloride has antidepressant activity and is used for research into cognitive and emotional disturbances related to cerebrovascular disease.
  • Potent, selective, and competitive inhibitor of monoamine oxidase A (MAO-A)
  • Inhibits MAO-B noncompetitively
  • Antidepressant activity
  • Used for research into cognitive and emotional disturbances related to cerebrovascular disease