Learn More
Medchemexpress LLC HY-119687 100mg Medchemexpress, Bifenazate CAS:149877-41-8 Purity:>98%
SDP
Catalog No. 501932541
Encompass_Preferred
Medchemexpress, HY-119687 100mg Bifenazate CAS:149877-41-8 Bifenazate is a carbazate acaricide that control 100% of mites at a concentration of 25 ppm [1] . Bifenazate is a positive allosteric modulator of GABA receptor [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2541 Supplier Medchemexpress LLC Supplier No. HY119687100MG
Medchemexpress, HY-119687 100mg Bifenazate CAS:149877-41-8 Bifenazate is a carbazate acaricide that control 100% of mites at a concentration of 25 ppm [1] . Bifenazate is a positive allosteric modulator of GABA receptor [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.