Learn More
AdipoGen Bifenazate
SDP
Supplier:  AdipoGen CDXB0122M100
Encompass_Preferred
Chemical. CAS 149877-41-8. Formula C17H20N2O3. MW 300.35. Synthetic. Bifenazate is a contact acaricide active against adult, eggs and nymphal life stages of spider, red and grass mites. It has a rapid knockdown effect usually less than 3 days and residual activity on the leaf lasting up to 4 weeks. It is suggested to act as a GABA gamma-aminobutyric acid antagonist in the peripheral nervous system at the neuromuscular synapse in insects. It blocks the GABA-activated chloride channels, resulting in overexcitation of the peripheral nervous systems of susceptible pests. This mode of action is reported to be unique among acaricides, which suggests the product could play an important future role in mite resistance management strategies. It is safe to sensitive flower parts and does not cause phyto-toxicity. Can be used as a reference compound.
Catalog No. 50-256-1673