Learn More
Medchemexpress LLC HY-W018791 100mg Medchemexpress, Bifendate CAS:73536-69-3 Purity:>98%
SDP
Catalog No. 502083431
Encompass_Preferred
Medchemexpress, HY-W018791 100mg Bifendate CAS:73536-69-3 Bifendate (DDB) is a synthetic intermediate of Schisandrin C with anti-HBV efficacy in research of chronic hepatitis B. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3431 Supplier Medchemexpress LLC Supplier No. HYW018791100MG
Add to Cart
Add to Cart
Medchemexpress, HY-W018791 100mg Bifendate CAS:73536-69-3 Bifendate (DDB) is a synthetic intermediate of Schisandrin C with anti-HBV efficacy in research of chronic hepatitis B. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.