Learn More
Medchemexpress LLC Bifenox | 42576-02-3 | 99.8% | 342.13 | 250 MG
SDP
Catalog No. 5000351850
Encompass_Preferred
Bifenox is a nitrophenyl ether herbicide that disrupts cellular membranes, inhibits photosynthesis, and inhibits protoporphyrinogen oxidase. It also increases ROS production in the microalgae Chlamydomonas reinhardtii.
  • Disrupts cellular membranes
  • Inhibits photosynthesis
  • Inhibits protoporphyrinogen oxidase
  • Increases ROS production in Chlamydomonas reinhardtii
  • Stable under recommended storage conditions
  • Recommended storage at room temperature for 3 years
  • Recommended storage in solvent at -80°C for 2 years
  • Recommended storage in solvent at -20°C for 1 year
Catalog No. 50-003-51850 Supplier Medchemexpress LLC Supplier No. 250MGHY136513
Bifenox is a nitrophenyl ether herbicide that disrupts cellular membranes, inhibits photosynthesis, and inhibits protoporphyrinogen oxidase. It also increases ROS production in the microalgae Chlamydomonas reinhardtii.
  • Disrupts cellular membranes
  • Inhibits photosynthesis
  • Inhibits protoporphyrinogen oxidase
  • Increases ROS production in Chlamydomonas reinhardtii
  • Stable under recommended storage conditions
  • Recommended storage at room temperature for 3 years
  • Recommended storage in solvent at -80°C for 2 years
  • Recommended storage in solvent at -20°C for 1 year