Learn More
Crescent Chemical Co Inc BIFENTHRIN 100 MG
SDP
Supplier:  Crescent Chemical Co Inc PSC10584000
Encompass_Preferred
Bifenthrin Insecticide (WS) CAS 82657-04-3 Molecular Weight - 422.8678 Storage - -18°C Catalog Num - DRE-C10584000 pesticide
Catalog No. 50-727-02