Learn More
Chem Service Inc Bifenthrin,82657-04-3,100MG
SDP
Catalog No. NC9199414
Encompass_Preferred
Molecular Formula C23H22ClF3O2, Purity 98.00%, Molecular Weight 422.87, Melting Point 69 °C, Synonyms: Capture, Talstar; Bifenthrin is chemically classified as a pyrethroid. It is a waxy solid, with a color ranging from an off-white to a pale tan, with a slightly sweet odor. It is mostly insoluble in water.
Catalog No. NC9199414 Supplier Chem Service Inc Supplier No. N11203100MG
Molecular Formula C23H22ClF3O2, Purity 98.00%, Molecular Weight 422.87, Melting Point 69 °C, Synonyms: Capture, Talstar; Bifenthrin is chemically classified as a pyrethroid. It is a waxy solid, with a color ranging from an off-white to a pale tan, with a slightly sweet odor. It is mostly insoluble in water.