Learn More
Medchemexpress LLC Bifenthrin | 82657-04-3 | 99.8% | 50 MG
SDP
Catalog No. 5000408723
Encompass_Preferred
Bifenthrin is a synthetic pyrethroid insecticide that prolongs the opening time of Nav1.8 sodium channels, leading to membrane depolarization and conductance block in the insect nervous system, which disrupts neural function.
  • Effective against *A. gambiae* and *C. quinquefasciatus*
  • Demonstrates good lethality against susceptible and resistant mosquitoes
  • Effectively inhibits blood sucking
  • Suitable for development as a mosquito-removal netting material
Catalog No. 50-004-08723 Supplier Medchemexpress LLC Supplier No. HYB082450MG
Bifenthrin is a synthetic pyrethroid insecticide that prolongs the opening time of Nav1.8 sodium channels, leading to membrane depolarization and conductance block in the insect nervous system, which disrupts neural function.
  • Effective against *A. gambiae* and *C. quinquefasciatus*
  • Demonstrates good lethality against susceptible and resistant mosquitoes
  • Effectively inhibits blood sucking
  • Suitable for development as a mosquito-removal netting material