Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bifeprunox mesylate | 350992-13-1 | MFCD09840052 | 99.9% | 481.56 | C25H27N3O5S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY14547A5MG

Encompass_Preferred

Bifeprunox mesylate is a research chemical that functions as a dopamine D2-like and 5-HT1A receptor partial agonist. Supplied as the mesylate salt for laboratory use, it is intended for preclinical in vitro and in vivo studies and is not for human or clinical application.

  • Acts as a dopamine D2-like and 5-HT1A receptor partial agonist.
  • Provided as the mesylate salt (solid form).
  • High reported purity (about 99.9% by HPLC).
  • Suitable for in vitro and in vivo preclinical research.
  • Available in small research pack sizes, such as 5 mg.
  • Molecular weight approximately 481.56 g/mol.

Catalog No. 50-002-21536


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.