Learn More
ATCC Bifidobacterium bifidum
Catalog No. 502381766
Encompass
Bifidobacterium bifidum strain S28a (Variant b) is a whole-genome sequenced bacterium that was isolated from the intestine of a human infant.
Catalog No. 50-238-1766 Supplier ATCC Supplier No. 15696
Bifidobacterium bifidum strain S28a (Variant b) is a whole-genome sequenced bacterium that was isolated from the intestine of a human infant.