Learn More
ATCC Bifidobacterium; longum; subsp. infantis
Catalog No. 502384961
Encompass
Bifidobacterium longum subsp. infantis strain S12 is a whole genome-sequenced type strain that was isolated from the intestine of a human infant. This strain has applications in bioinformatics and as a bacteriophage host.
Catalog No. 50-238-4961 Supplier ATCC Supplier No. 15697
Bifidobacterium longum subsp. infantis strain S12 is a whole genome-sequenced type strain that was isolated from the intestine of a human infant. This strain has applications in bioinformatics and as a bacteriophage host.