Learn More
ATCC Bifidobacterium longum; subsp. longum
Supplier:  ATCC 15707
Encompass
Bifidobacterium longum subsp. longum strain E194b (Variant a) is a whole genome-sequenced type strain that was isolated from the intestine of an adult.
Catalog No. 50-238-2519