Learn More
Biolog Inc BIFIDO - Bifidobacterium Selective Agar, 4 Plates/Pkg
SDP
Catalog No. NC1268131
Encompass
This item has a minimum qty of 25 per supplier requirements.
Bifidobacterium Selective Agar (BIFIDO) is a selective media for the isolation and enumeration of Bifidobacterium species. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.BIFIDO agar contains a reinforced clostridial agar formulation as the basal nutritive media. Selective and differential compounds include iodoacetic acid and 2, 3, 5-triphenyltetrazolium chloride (TTC). Iodoacetate inhibits glyceraldehyde-3-phosphate dehydrogenase, drastically reducing the growth of non-bifidobacterial colonies. TTC makes it possible to differentiate between other species and Bifidobacterium, since the bifidobacterial develop in large white colonies.
Catalog No. NC1268131 Supplier Biolog Inc Supplier No. AS6423
This item has a minimum qty of 25 per supplier requirements.
Bifidobacterium Selective Agar (BIFIDO) is a selective media for the isolation and enumeration of Bifidobacterium species. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.BIFIDO agar contains a reinforced clostridial agar formulation as the basal nutritive media. Selective and differential compounds include iodoacetic acid and 2, 3, 5-triphenyltetrazolium chloride (TTC). Iodoacetate inhibits glyceraldehyde-3-phosphate dehydrogenase, drastically reducing the growth of non-bifidobacterial colonies. TTC makes it possible to differentiate between other species and Bifidobacterium, since the bifidobacterial develop in large white colonies.