Learn More
Medchemexpress LLC Bifluranol (BX341) | 34633-34-6 | 292.32 | 50 MG
SDP
Catalog No. 5000445756
Encompass_Preferred
Bifluranol (BX341) is an anti-androgen compound intended for research use only. Studies have demonstrated its absorption, distribution, and excretion profiles in various animal models, including mice, rats, ferrets, and dogs. It is readily absorbed orally, but blood concentrations remain low due to significant hepatic uptake and biliary excretion.
  • Acts as an androgen receptor inhibitor.
  • Readily absorbed after oral administration.
  • Exhibits low blood concentrations due to hepatic uptake and biliary excretion.
  • Suitable for research applications.
Catalog No. 50-004-45756 Supplier Medchemexpress LLC Supplier No. HYU0022950MG
Bifluranol (BX341) is an anti-androgen compound intended for research use only. Studies have demonstrated its absorption, distribution, and excretion profiles in various animal models, including mice, rats, ferrets, and dogs. It is readily absorbed orally, but blood concentrations remain low due to significant hepatic uptake and biliary excretion.
  • Acts as an androgen receptor inhibitor.
  • Readily absorbed after oral administration.
  • Exhibits low blood concentrations due to hepatic uptake and biliary excretion.
  • Suitable for research applications.