Learn More
Bollé Safety™ Bifocal D28 Arizona Rx Lens
Catalog No. 18072205
Change view
Click to view available options
Lens Material:
CR39
Polycarbonate
Trivex
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Material
18-072-205 CR39
18-072-158 Polycarbonate
18-072-180 Trivex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-072-205 Supplier Bollé Safety™ Supplier No. CR39UV1
This item is not returnable. View return policy
Bifocal lens option for prescription safety glasses.
  • For use with prescription frames
Specifications
Lens Coating Uncoated
Lens Type Bifocal
Product Type Prescription Lens
Disposable Reusable
Lens Material CR39
Autoclavable Not Autoclavable