Learn More
Apexbio Technology LLC Bifonazole 60628-96-8 1g
SDP
Catalog No. 5000089974
Encompass_Preferred
Bifonazole (60628-96-8) is a small-molecule inhibitor targeting sterol 14 -demethylase It is designed to block this enzyme thereby impairing ergosterol biosynthesis critical to fungal membrane structure and function Bifonazole exerts its biological activity primarily through inhibition of sterol 14 -demethylase In in vitro studies Bifonazole demonstrates antifungal inhibitory activity with reported IC50 values generally ranging from 0 5 to 4 g/mL depending on fungal species and experimental conditions Based on these pharmacological properties Bifonazole holds research potential in antifungal efficacy evaluation and mechanism-specific assays targeting sterol biosynthesis and membrane disruption
Catalog No. 50-000-89974 Supplier Apexbio Technology LLC Supplier No. B18971000
Bifonazole (60628-96-8) is a small-molecule inhibitor targeting sterol 14 -demethylase It is designed to block this enzyme thereby impairing ergosterol biosynthesis critical to fungal membrane structure and function Bifonazole exerts its biological activity primarily through inhibition of sterol 14 -demethylase In in vitro studies Bifonazole demonstrates antifungal inhibitory activity with reported IC50 values generally ranging from 0 5 to 4 g/mL depending on fungal species and experimental conditions Based on these pharmacological properties Bifonazole holds research potential in antifungal efficacy evaluation and mechanism-specific assays targeting sterol biosynthesis and membrane disruption