Learn More
Medchemexpress LLC Bifonazole (Standard) | 60628-96-8 | 99.4% | 250 MG
SDP
Catalog No. 5000407062
Encompass_Preferred
Bifonazole (Standard) is the analytical standard of Bifonazole. It is intended for research and analytical applications. Bifonazole (Bay H-4502) is an imidazole antifungal agent. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Bifonazole
  • Intended for research and analytical applications
  • Imidazole antifungal agent
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
Catalog No. 50-004-07062 Supplier Medchemexpress LLC Supplier No. 250MGHYB0301R
Bifonazole (Standard) is the analytical standard of Bifonazole. It is intended for research and analytical applications. Bifonazole (Bay H-4502) is an imidazole antifungal agent. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Bifonazole
  • Intended for research and analytical applications
  • Imidazole antifungal agent
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS