Learn More
World Precision Instrument Bifurcated Fiber Optic Assemblies-BIF44
SDP
Catalog No. 502538916
Encompass
Various bifurcated fibers for customized applications such as: Reference measurement in dual channel spectrometers, Combine UV and VIS light sources, and Couple light into two sample cells.
Catalog No. 50-253-8916 Supplier World Precision Instrument Supplier No. BIF44
Various bifurcated fibers for customized applications such as: Reference measurement in dual channel spectrometers, Combine UV and VIS light sources, and Couple light into two sample cells.