Learn More
Waters Corp Waters Corporation Calibration Stock Standard, 125 mL, Four Sweeteners and Preservatives, Certified Accuracy
Catalog No. 502038289 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This calibration stock standard is a 125 mL solution of four artificial sweeteners and preservatives (caffeine, benzoate, sorbate, and acesulfame potassium), each at 1,000 mg/L in an acetate buffer reagent with 5% ethanol. Analytes are traceable via USP reference standards to SI units and meet ISO Guide 31. Designed for QC methods employing HPLC, UPLC, or UHPLC.
  • Ready-to-use solution for efficient preparation.
  • Certified accuracy within 2% for precise LC analyses.
  • One-year shelf life when stored properly.
  • Versatile standard for food, beverage, and pharmaceutical sectors.
  • Supports quantitative and qualitative analysis of sweeteners and preservatives.
  • Aids in troubleshooting LC instruments.
  • Traceable analytes meet ISO Guide 31.
  • Suitable for QC methods using HPLC, UPLC, or UHPLC.
  • Certificate of analysis included.
Catalog No. 50-203-8289 Supplier Waters Corp Supplier No. 186007980
This calibration stock standard is a 125 mL solution of four artificial sweeteners and preservatives (caffeine, benzoate, sorbate, and acesulfame potassium), each at 1,000 mg/L in an acetate buffer reagent with 5% ethanol. Analytes are traceable via USP reference standards to SI units and meet ISO Guide 31. Designed for QC methods employing HPLC, UPLC, or UHPLC.
  • Ready-to-use solution for efficient preparation.
  • Certified accuracy within 2% for precise LC analyses.
  • One-year shelf life when stored properly.
  • Versatile standard for food, beverage, and pharmaceutical sectors.
  • Supports quantitative and qualitative analysis of sweeteners and preservatives.
  • Aids in troubleshooting LC instruments.
  • Traceable analytes meet ISO Guide 31.
  • Suitable for QC methods using HPLC, UPLC, or UHPLC.
  • Certificate of analysis included.