Learn More
MSC Big Ass Fans Evaporative Cooler, 3,400 CFM, 2,300 sq ft, 50 Gallon
Catalog No. 5000507141
Encompass
The Cool-Space 350 Evaporative Cooler is a heavy-duty cooling solution for outdoor patios, warehouses, and large work areas. It drops temperatures by up to 33°F, covering 2,300 sq ft. Its 50-gallon reservoir provides 10 hours of operation, offering the longest runtime in its class. It is most effective in low humidity but still delivers substantial cooling in moderate environments.
  • 20-inch fan pushes 3,400 CFM through 8-inch cooling media for evaporative cooling.
  • Quiet operation at 69 dBA.
  • 50-gallon reservoir provides 10 hours of runtime, with option for continuous garden hose refill.
  • Low-water shutoff protects the pump.
  • Standard dial control includes a pump on/off switch.
  • Durable UV-resistant housing and algae-resistant GREENGUARD Gold-certified media.
  • Equipped with four swivel casters, including two locking ones.
Catalog No. 50-005-07141 Supplier MSC Supplier No. 89819015
The Cool-Space 350 Evaporative Cooler is a heavy-duty cooling solution for outdoor patios, warehouses, and large work areas. It drops temperatures by up to 33°F, covering 2,300 sq ft. Its 50-gallon reservoir provides 10 hours of operation, offering the longest runtime in its class. It is most effective in low humidity but still delivers substantial cooling in moderate environments.
  • 20-inch fan pushes 3,400 CFM through 8-inch cooling media for evaporative cooling.
  • Quiet operation at 69 dBA.
  • 50-gallon reservoir provides 10 hours of runtime, with option for continuous garden hose refill.
  • Low-water shutoff protects the pump.
  • Standard dial control includes a pump on/off switch.
  • Durable UV-resistant housing and algae-resistant GREENGUARD Gold-certified media.
  • Equipped with four swivel casters, including two locking ones.