Learn More
Bioworld BIG CHAP, 5 g
Supplier:  Bioworld 402300001
Encompass_Preferred
Big CHAP [N,N'-Bis(3-D-gluconamidoproply)cholamide]A non-ionic detergent analog of CHAPS and CHAPSOHigh Purity Grade Identification (IR)(P/F)- Pass Moisture(%)- 1.8 Reassay Date- ReportNon-ionic detergentConductivity: <25µS in a 10% solutionCritical micelle concentration: 3.4mM (25°C)Aggregation number: 10Appearance: Fine colorless crystlesBoiling Point : 1191.28 °C at 760 mmHg (Predicted)Density : 1.368Melting Point: 136-145°C136-145°C
Catalog No. 50-255-642