Learn More
Cole-Parmer Extech 445713 Big Digit Thermohygrometer with Remote Temperature Probe
Catalog No. 50992483 Shop All Cole-Parmer Products
Encompass
Designed to monitor relative humidity, and temperature. Features include a super large display, C/F switchable, Min/Max with reset function, low battery indication and a built-in stand for sitting on a desk and hardware for mounting on a wall.
Catalog No. 50-992-483 Supplier Cole-Parmer Supplier No. 3780915
Designed to monitor relative humidity, and temperature. Features include a super large display, C/F switchable, Min/Max with reset function, low battery indication and a built-in stand for sitting on a desk and hardware for mounting on a wall.