Learn More
Sparkfun Electronics BIG EASY DRIVER
Supplier:  Sparkfun Electronics ROB12859
Encompass_Preferred
Big Easy Driver. Stepper motor driver board for bi-polar stepper motors up to a max 2A/phase. It is based on the Allegro A4988 stepper driver chip. It’s the next version of the popular Easy Driver board. Bi-polar Microstepping Driver. 2A/Phase Max. 1.4-1.7A/Phase w/o Heatsink. Max Motor Drive Voltage: 30V. On-board 5V/3.3V Regulation.
Catalog No. NC0842804