Learn More
Echelon Biosciences Research Labs Big Endothelin 2 (1-37), human - 0.5 mg, CAS#: 132699-72-0
SDP
Supplier:  Echelon Biosciences Research Labs 331400.5MG
Encompass_Preferred
Big Endothelin 2 (1-37), human - 0.5mg , Cys-Ser-Cys-Ser-Ser-Trp-Leu-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-Val-Asn-Thr-Pro-Glu-Gln-Thr-Ala-Pro-Tyr-Gly-Leu-Gly-Asn-Pro-Pro-OH Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8378