Learn More
Enzo Life Sciences Big Endothelin-1 (human) (1-38) (0.1mg). CAS: 121014-53-7
SDP
Supplier:  Enzo Life Sciences ALX152001PC01
Encompass_Preferred
Biological precursor of endothelin 1. Cleaved to yield endothelin-1 via the activity of the endothelin-converting enzyme (ECE). Endothelins are endothelium-derived vasoconstrictor peptides. Alternative name: Big ET-1 (1-38). Formula: C189H282N48O56S5. MW: 4283.0. Purity: ≥98% (HPLC). Solubility: 1mg/ml soluble in water; also soluble in 0.5% acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-7780