Learn More
CPC Scientific Glp-Leu-Gly-Pro-Gln-Gly-Pro-Pro-His-Leu-Val-Ala-Asp-Pro-Ser-Lys-Lys-Gln-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GAST001B
Encompass
SEQUENCE: Glp-Leu-Gly-Pro-Gln-Gly-Pro-Pro-His-Leu-Val-Ala-Asp-Pro-Ser-Lys-Lys-Gln-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Glp-LGPQGPPHLVADPSKKQGPWLEEEEEAYGWMDF-NH2MOLECULAR FORMULA: C176H251N43O53S1MOLECULAR WEIGHT: 3849.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [60675-77-6]SYNONYMS: RESEARCH AREA: GastrointestinalREFERENCES:A.M. Choudhury et al., Hoppe-Seyler's Z. Physiol. Chem., 361, 1719 (1980)SKU(s): GAST-001A, GAST-001B
Catalog No. 50-210-9315