Learn More
Echelon Biosciences Research Labs Big Gastrin, sulfated - 0.5 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 471800.5MG
Encompass_Preferred
Big Gastrin, sulfated - 0.5mg , Glp-Leu-Gly-Pro-Gln-Gly-Pro-Pro-His-Leu-Val-Ala-Asp-Pro-Ser-Lys-Lys-Gln-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr(SO3H)-Gly-Trp-Met-Asp-Phe-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8512