Learn More
Bellco Glass, Inc. Big Mouth Amber S/T Vial,2.0mL 12x32mm - 10-425 Thd
SDP
Catalog No. 501945372
Encompass_Preferred
Clear vials are manufactured from type 1 class a Borosilicate glass. Amber vials are manufactured from type1 amber glass. Designed for use with varian and other auto-samplers. MADE IN THE USA. Size: 12x32mm Cap size: 10-425
Catalog No. 50-194-5372 Supplier Bellco Glass, Inc. Supplier No. 207000180
Clear vials are manufactured from type 1 class a Borosilicate glass. Amber vials are manufactured from type1 amber glass. Designed for use with varian and other auto-samplers. MADE IN THE USA. Size: 12x32mm Cap size: 10-425