Learn More
Bellco Glass, Inc. Big Mouth Amber Vial, 2.0mL Crimp Top - 12x32mm
SDP
Catalog No. 501945359
Encompass_Preferred
This 11mm crimp vial is made of amber borosilicate glass with a 40% larger opening that prevents broken needles due to increased target area. Designed to work in Agilent and other autosamplers. Sample capacity of 2mL, 12mm in diameter, and 32mm in height. Vials compatible with 11mm crimp and Poly Crimp Seals. 1,000 vials per case with 10 shelf packs of 100 vials packed in plastic shrink wrapped scallop packaging. MADE IN THE USA.
Catalog No. 50-194-5359 Supplier Bellco Glass, Inc. Supplier No. 207000060
This 11mm crimp vial is made of amber borosilicate glass with a 40% larger opening that prevents broken needles due to increased target area. Designed to work in Agilent and other autosamplers. Sample capacity of 2mL, 12mm in diameter, and 32mm in height. Vials compatible with 11mm crimp and Poly Crimp Seals. 1,000 vials per case with 10 shelf packs of 100 vials packed in plastic shrink wrapped scallop packaging. MADE IN THE USA.